Assay Details
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025911.2 | NP_080187.2 | 2 - 3 | 232 | 67 | |
| GenBank mRNA | AK007917.1 | - | 2 - 3 | 233 | 67 | |
| BC006734.1 | - | 2 - 3 | 137 | 3156328 | 67 | |
| BC049073.1 | - | 2 - 3 | 201 | 5707404 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007917.1, BC006734.1, BC049073.1, NM_025911.2 transcripts.
Set Membership: |
|

Ccdc91
Genomic Map