Assay Details
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025911.2 | NP_080187.2 | 4 - 5 | 471 | 58 | |
| GenBank mRNA | AK007017.1 | - | 3 - 4 | 395 | 58 | |
| BC006734.1 | - | 4 - 5 | 376 | 3156328 | 58 | |
| BC049073.1 | - | 4 - 5 | 440 | 5707404 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc91
Genomic Map