Assay Details
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025911.2 | NP_080187.2 | 7 - 8 | 865 | 67 | |
| GenBank mRNA | AK007917.1 | - | 7 - 8 | 866 | 67 | |
| BC006734.1 | - | 7 - 8 | 770 | 3156328 | 67 | |
| BC024946.1 | - | 2 - 3 | 123 | 4010938 | 67 | |
| BC049073.1 | - | 7 - 8 | 834 | 5707404 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc91
Genomic Map