Assay Details
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025911.2 | NP_080187.2 | 8 - 9 | 969 | 59 | |
| GenBank mRNA | AK007917.1 | - | 8 - 9 | 970 | 59 | |
| BC006734.1 | - | 8 - 9 | 874 | 3156328 | 59 | |
| BC024946.1 | - | 3 - 4 | 227 | 4010938 | 59 | |
| BC049073.1 | - | 8 - 9 | 938 | 5707404 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc91
Genomic Map