Assay Details
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025911.2 | NP_080187.2 | 9 - 10 | 1057 | 94 | |
| GenBank mRNA | AK007917.1 | - | 9 - 10 | 1058 | 94 | |
| BC006734.1 | - | 9 - 10 | 962 | 3156328 | 94 | |
| BC024946.1 | - | 4 - 5 | 315 | 4010938 | 94 | |
| BC049073.1 | - | 9 - 10 | 1026 | 5707404 | 94 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc91
Genomic Map