| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400008534.mm.1 |
NM_001048189.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008534.mm.1 |
NM_001284190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008534.mm.1 |
NM_030231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008534.mm.1 |
TC0400001180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400008533.mm.1 |
NM_001048189.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008533.mm.1 |
NM_001284190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008533.mm.1 |
NM_030231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008533.mm.1 |
TC0400001180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400008542.mm.1 |
NM_001048189.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008542.mm.1 |
NM_001284190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008542.mm.1 |
NM_030231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008542.mm.1 |
TC0400001180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400008538.mm.1 |
NM_001048189.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008538.mm.1 |
NM_001284190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008538.mm.1 |
NM_030231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008538.mm.1 |
TC0400001180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400008539.mm.1 |
NM_001048189.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008539.mm.1 |
NM_001284190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008539.mm.1 |
NM_030231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008539.mm.1 |
TC0400001180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400008535.mm.1 |
NM_001048189.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008535.mm.1 |
NM_001284190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008535.mm.1 |
NM_030231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008535.mm.1 |
TC0400001180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400008536.mm.1 |
NM_001048189.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008536.mm.1 |
NM_001284190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008536.mm.1 |
NM_030231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008536.mm.1 |
TC0400001180.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400008541.mm.1 |
NM_001048189.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008541.mm.1 |
NM_001284190.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008541.mm.1 |
NM_030231.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400008541.mm.1 |
TC0400001180.mm.1 |
MTA TC |