Assay Details
Gene Symbol: Agbl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048189.4 | NP_001041654.2 | 5 - 6 | 768 | 60 | |
NM_001284190.1 | NP_001271119.1 | 5 - 6 | 768 | 60 | ||
NM_030231.3 | NP_084507.1 | 5 - 6 | 768 | 60 | ||
GenBank mRNA | AK016502.1 | - | 5 - 6 | 733 | 60 | |
DQ867039.1 | - | 5 - 6 | 675 | 60 | ||
DQ867040.1 | - | 5 - 6 | 675 | 60 | ||
FN429928.1 | - | 5 - 6 | 594 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
