Assay Details
| Gene Symbol: Cyp2j5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010007.4 | NP_034137.1 | 2 - 3 | 482 | 76 | |
| GenBank mRNA | AK098119.1 | - | 2 - 3 | 413 | 76 | |
| AK143932.1 | - | 2 - 3 | 483 | 76 | ||
| AK165404.1 | - | 2 - 3 | 492 | 76 | ||
| BC021624.1 | - | 2 - 3 | 449 | 4161995 | 76 | |
| U62294.1 | - | 2 - 3 | 414 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2j5
Genomic Map