Assay Details
| Gene Symbol: Cyp2j5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010007.4 | NP_034137.1 | 5 - 6 | 969 | 97 | |
| GenBank mRNA | AK098119.1 | - | 5 - 6 | 900 | 97 | |
| AK143932.1 | - | 5 - 6 | 970 | 97 | ||
| AK149552.1 | - | 5 - 6 | 936 | 97 | ||
| AK165404.1 | - | 5 - 6 | 979 | 97 | ||
| BC021624.1 | - | 5 - 6 | 936 | 4161995 | 97 | |
| U62294.1 | - | 5 - 6 | 901 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp2j5
Genomic Map