Assay Details
| Gene Symbol: G2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015099.2 | NP_001015099.2 | 11 - 12 | 1531 | 105 | |
| NM_001167963.1 | NP_001161435.1 | 12 - 13 | 1600 | 105 | ||
| GenBank mRNA | AK033513.1 | - | 12 - 13 | 1542 | 105 | |
| AK129332.1 | - | 11 - 12 | 1434 | 105 | ||
| BC086455.1 | - | 11 - 12 | 1404 | 30458160 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

G2e3
Genomic Map