Assay Details
| Gene Symbol: Efcab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145549.1 | NP_663524.1 | 1756 | 163 | ||
| GenBank mRNA | AK142188.1 | - | 10 - 11 | 1961 | 163 | |
| AK220422.1 | - | 1705 | 163 | |||
| BC020077.1 | - | 1756 | 4018470 | 163 | ||
| BC112327.1 | - | 10 - 11 | 1607 | 5362489 | 163 | |
More Information
Set Membership: |
|

Efcab7
Genomic Map