Assay Details
| Gene Symbol: Efcab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145549.1 | NP_663524.1 | 1873 | 121 | ||
| GenBank mRNA | AK142188.1 | - | 11 - 12 | 2078 | 121 | |
| AK220422.1 | - | 1822 | 121 | |||
| BC020077.1 | - | 1873 | 4018470 | 121 | ||
| BC112327.1 | - | 11 - 12 | 1724 | 5362489 | 121 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23146396 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184304 | BC020077.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184304 | NM_145549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015312 | BC020077.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015312 | BC112327.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015312 | AK142188.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015312 | AK143777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2015312 | AK220422.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918141 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 882383 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676177 | NM_011282.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676177 | U15443.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676177 | X81650.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648950 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342620 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174890 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39925 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267738 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11202 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88791 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170634 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015312 | BC020077.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015312 | BC112327.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015312 | AK142188.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015312 | AK143777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2015312 | AK220422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184304 | BC020077.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184304 | NM_145549.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23146401 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170634 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05333019 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05333020 | - | - |
| Whole Mouse Genome G4122A | A_51_P485275 | BC020077.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485275 | BC112327.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485275 | AK142188.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485275 | AK220422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P485275 | NM_145549.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117329 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101821 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111492 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125717 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170634 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05333019 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05333020 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03292012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03595408 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03595403 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03261036 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03595405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03420111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03773264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03344971 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03595406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03773267 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03595412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23146396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23146487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03990104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03420564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04033972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03491070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03595403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03595412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03595425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04018749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23146474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03595405 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10506182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506173 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506185 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506181 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506176 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10506179 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451349_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10506170 | - | - |
| Mouse Genome 430 2.0 Array | 1451349_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451349_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230500_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230500_at | - | - |
| Mouse Expression Set 430 | 1451349_at | - | - |
| Mouse Expression Set 430 | 1451349_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451349_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451349_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451349_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451349_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 695705 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4150537 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4150537 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004150537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2768638 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2768638 | - | - |
| mouse-6 v1.1 (Illumina) | scl25242.12.1_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4150537 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25242.12.1_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150537 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2768638 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2768638 | - | - |

Efcab7
Genomic Map