Assay Details
| Gene Symbol: Efcab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145549.1 | NP_663524.1 | 58 | 69 | ||
| GenBank mRNA | AK143777.1 | - | 116 | 69 | ||
| BC020077.1 | - | 58 | 4018470 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143777.1, BC020077.1, NM_145549.1 transcripts.
Set Membership: |
|

Efcab7
Genomic Map