Assay Details
| Gene Symbol: Efcab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145549.1 | NP_663524.1 | 1264 | 115 | ||
| GenBank mRNA | AK142188.1 | - | 7 - 8 | 1469 | 115 | |
| AK220422.1 | - | 1213 | 115 | |||
| BC020077.1 | - | 1264 | 4018470 | 115 | ||
| BC112327.1 | - | 7 - 8 | 1115 | 5362489 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Efcab7
Genomic Map