Assay Details
| Gene Symbol: Sepw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009156.2 | NP_033182.1 | 4 - 5 | 275 | 70 | |
| GenBank mRNA | AF015284.1 | - | 4 - 5 | 230 | 70 | |
| AF241527.2 | - | 4 - 5 | 228 | 70 | ||
| AK002870.1 | - | 4 - 5 | 251 | 70 | ||
| AK028124.1 | - | 4 - 5 | 245 | 70 | ||
| BC052719.1 | - | 4 - 5 | 237 | 6814242 | 70 | |
| BU604713.1 | - | 4 - 5 | 308 | 70 | ||
| U67890.1 | - | 4 - 5 | 227 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sepw1
Genomic Map