Assay Details
| Gene Symbol: Slc27a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011978.2 | NP_036108.2 | 2 - 3 | 819 | 67 | |
| GenBank mRNA | AF033031.1 | - | 2 - 3 | 700 | 67 | |
| AJ223958.1 | - | 2 - 3 | 737 | 67 | ||
| AK144069.1 | - | 2 - 3 | 821 | 67 | ||
| AK144070.1 | - | 2 - 3 | 733 | 67 | ||
| AK165446.1 | - | 2 - 3 | 820 | 67 | ||
| BC013442.1 | - | 2 - 3 | 802 | 4236616 | 67 | |
| BC022170.1 | - | 2 - 3 | 798 | 4159081 | 67 | |
| BC024735.1 | - | 2 - 3 | 788 | 5123650 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a2
Genomic Map