Assay Details
| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146010.1 | NP_001139482.1 | 11 - 12 | 1282 | 93 | |
| NM_199012.2 | NP_950177.2 | 12 - 13 | 1354 | 93 | ||
| GenBank mRNA | AK122368.1 | - | 12 - 13 | 1246 | 93 | |
| AK141949.1 | - | 11 - 12 | 1282 | 93 | ||
| BC060647.1 | - | 12 - 13 | 1316 | 6854041 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fchsd2
Genomic Map