Assay Details
| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146010.1 | NP_001139482.1 | 14 - 15 | 1693 | 73 | |
| NM_199012.2 | NP_950177.2 | 15 - 16 | 1765 | 73 | ||
| GenBank mRNA | AK122368.1 | - | 15 - 16 | 1657 | 73 | |
| AK140322.1 | - | 14 - 15 | 1672 | 73 | ||
| AK141949.1 | - | 14 - 15 | 1693 | 73 | ||
| BC034086.1 | - | 2 - 3 | 240 | 73 | ||
| BC039788.1 | - | 1 - 2 | 167 | 73 | ||
| BC060647.1 | - | 15 - 16 | 1727 | 6854041 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fchsd2
Genomic Map