Assay Details
| Gene Symbol: Fchsd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146010.1 | NP_001139482.1 | 8 - 9 | 941 | 70 | |
| NM_199012.2 | NP_950177.2 | 8 - 9 | 941 | 70 | ||
| GenBank mRNA | AK122368.1 | - | 8 - 9 | 833 | 70 | |
| AK140322.1 | - | 8 - 9 | 920 | 70 | ||
| AK141949.1 | - | 8 - 9 | 941 | 70 | ||
| AK157054.1 | - | 8 - 9 | 914 | 70 | ||
| BC060647.1 | - | 8 - 9 | 903 | 6854041 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fchsd2
Genomic Map