Assay Details
| Gene Symbol: Foxp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110824.1 | NP_001104294.1 | 7 - 8 | 1334 | 75 | |
| NM_001110825.1 | NP_001104295.1 | 7 - 8 | 1334 | 75 | ||
| NM_028767.2 | NP_083043.2 | 7 - 8 | 1331 | 75 | ||
| GenBank mRNA | AB052766.1 | - | 7 - 8 | 1165 | 75 | |
| AK004693.1 | - | 7 - 8 | 1142 | 75 | ||
| AK143194.1 | - | 7 - 8 | 1181 | 75 | ||
| AY135029.1 | - | 7 - 8 | 1122 | 75 | ||
| BC043702.1 | - | 7 - 8 | 1108 | 5351757 | 75 | |
| BC052407.1 | - | 7 - 8 | 1137 | 5716170 | 75 | |
| BC057110.1 | - | 7 - 8 | 1331 | 6417088 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Foxp4
Genomic Map