Assay Details
| Gene Symbol: Wdr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159527.1 | NP_001152999.1 | 17 - 18 | 2076 | 134 | |
| NM_172470.3 | NP_766058.3 | 18 - 19 | 2109 | 134 | ||
| GenBank mRNA | AK029896.1 | - | 17 - 18 | 2031 | 134 | |
| AK083971.1 | - | 17 - 18 | 2021 | 134 | ||
| AK173151.1 | - | 18 - 19 | 2051 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr35
Genomic Map