Assay Details
| Gene Symbol: Wdr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159527.1 | NP_001152999.1 | 18 - 19 | 2220 | 73 | |
| NM_172470.3 | NP_766058.3 | 19 - 20 | 2253 | 73 | ||
| GenBank mRNA | AK029896.1 | - | 18 - 19 | 2175 | 73 | |
| AK083971.1 | - | 18 - 19 | 2165 | 73 | ||
| AK173151.1 | - | 19 - 20 | 2195 | 73 | ||
| BC056925.1 | - | 19 - 20 | 2362 | 5704780 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr35
Genomic Map