Assay Details
| Gene Symbol: Nol10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008421.1 | NP_001008421.1 | 6 - 7 | 524 | 80 | |
| GenBank mRNA | AK148210.1 | - | 6 - 7 | 540 | 80 | |
| AK160864.1 | - | 6 - 7 | 560 | 80 | ||
| AK164400.1 | - | 6 - 7 | 550 | 80 | ||
| AK166160.1 | - | 6 - 7 | 553 | 80 | ||
| BC086676.1 | - | 6 - 7 | 524 | 30659737 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nol10
Genomic Map