Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200037660.mm.1 |
NM_001024856.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037660.mm.1 |
NM_001024856.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037660.mm.1 |
NM_001024857.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037660.mm.1 |
NM_001024857.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037660.mm.1 |
NM_080788.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037660.mm.1 |
NM_080788.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037660.mm.1 |
TC0200005502.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037659.mm.1 |
NM_001024856.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037659.mm.1 |
NM_001024856.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037659.mm.1 |
NM_001024857.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037659.mm.1 |
NM_001024857.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037659.mm.1 |
NM_080788.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037659.mm.1 |
NM_080788.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037659.mm.1 |
TC0200005502.mm.1 |
MTA TC |