Assay Details
| Gene Symbol: Ttc30b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028235.1 | NP_082511.1 | 1 | 2414 | 137 | |
| GenBank mRNA | AK011097.1 | - | 1 | 2391 | 137 | |
| AK147134.1 | - | 1 | 2364 | 137 | ||
| BC026507.1 | - | 1 | 1606 | 137 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | BC151184.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | BC157929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | BC026507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | AK147134.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | BC151060.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | BC151184.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | BC157929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | BC026507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | AK147134.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P96748 | BC151060.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275033 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146868 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | BC151184.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | BC157929.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | BC026507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | AK147134.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | BC151060.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | BC151184.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | BC157929.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | BC026507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | AK147134.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P96748 | BC151060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223891 | BC151184.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223891 | BC157929.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223891 | BC026507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223891 | AK147134.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P223891 | BC151060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | BC151184.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | BC157929.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | BC026507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | AK147134.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | BC151060.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | BC151184.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | BC157929.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | BC026507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | AK147134.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P96748 | BC151060.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21347032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21346947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01463956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01559785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01260692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01901440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01559783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01693308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01901442 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01596836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01693304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01559778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21346970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21347021 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10483823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10483821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10483820 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423672_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10483819 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10483822 | - | - |
| Mouse Genome 430 2.0 Array | 1423672_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1423672_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72421_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72421_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200032425.mm.1 | NM_028235.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032425.mm.1 | NM_028235.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032425.mm.1 | TC0200003798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200032427.mm.1 | NM_028235.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032427.mm.1 | NM_028235.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032427.mm.1 | TC0200003798.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423672_PM_at | - | - |
| Mouse Expression Set 430 | 1423672_at | - | - |
| Murine Genome U74v2 Set | 160908_r_at | - | - |
| Murine Genome U74v2 Set | 160908_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423672_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423672_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423672_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423672_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 353985 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247043 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2804996 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2804996 | - | - |

Ttc30b
Genomic Map