Assay Details
| Gene Symbol: Frzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011356.4 | NP_035486.1 | 3 - 4 | 1151 | 92 | |
| GenBank mRNA | AK019093.2 | - | 3 - 4 | 822 | 92 | |
| AK029941.1 | - | 3 - 4 | 1153 | 92 | ||
| BC016884.1 | - | 3 - 4 | 998 | 4237554 | 92 | |
| U68058.1 | - | 3 - 4 | 607 | 92 | ||
| U88568.1 | - | 3 - 4 | 959 | 92 | ||
| U91905.1 | - | 3 - 4 | 1393 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frzb
Genomic Map