Assay Details
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010806.1 | NP_034936.1 | 12 - 13 | 1781 | 59 | |
GenBank mRNA | AK016557.1 | - | 11 - 12 | 1900 | 59 | |
CK637291.1 | - | 3 - 4 | 433 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
