Assay Details
| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010806.1 | NP_034936.1 | 14 - 15 | 1966 | 127 | |
| GenBank mRNA | AK016557.1 | - | 13 - 14 | 2085 | 127 | |
| CK637291.1 | - | 5 - 6 | 618 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mllt4
Genomic Map