Assay Details
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010806.1 | NP_034936.1 | 17 - 18 | 2413 | 66 | |
GenBank mRNA | AK016557.1 | - | 16 - 17 | 2532 | 66 | |
CN537179.1 | - | 3 - 4 | 283 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
