Assay Details
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010806.1 | NP_034936.1 | 21 - 22 | 3127 | 122 | |
GenBank mRNA | CN538841.1 | - | 3 - 4 | 348 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
