Assay Details
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010806.1 | NP_034936.1 | 25 - 26 | 3469 | 69 | |
GenBank mRNA | BU611478.1 | - | 1 - 2 | 71 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
