| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700001233.mm.1 |
NM_010806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001233.mm.1 |
NM_010806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001233.mm.1 |
TC1700000159.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001229.mm.1 |
NM_010806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001229.mm.1 |
NM_010806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001229.mm.1 |
TC1700000159.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001231.mm.1 |
NM_010806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001231.mm.1 |
NM_010806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001231.mm.1 |
TC1700000159.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001225.mm.1 |
NM_010806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001225.mm.1 |
NM_010806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001225.mm.1 |
TC1700000159.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001224.mm.1 |
NM_010806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001224.mm.1 |
NM_010806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001224.mm.1 |
TC1700000159.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001227.mm.1 |
NM_010806.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001227.mm.1 |
NM_010806.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001227.mm.1 |
TC1700000159.mm.1 |
MTA TC |