Assay Details
| Gene Symbol: Acss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080575.2 | NP_542142.1 | 11 - 12 | 1724 | 105 | |
| GenBank mRNA | AB046742.1 | - | 11 - 12 | 1713 | 105 | |
| AK088244.1 | - | 11 - 12 | 1703 | 105 | ||
| AK146568.1 | - | 11 - 12 | 1725 | 105 | ||
| AK158119.1 | - | 11 - 12 | 1719 | 105 | ||
| AK172412.1 | - | 11 - 12 | 1710 | 105 | ||
| AK172470.1 | - | 11 - 12 | 1699 | 105 | ||
| AK173275.1 | - | 11 - 12 | 1685 | 105 | ||
| BC030930.1 | - | 7 - 8 | 898 | 4911679 | 105 | |
| BC092278.1 | - | 11 - 12 | 1753 | 3710505 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acss1
Genomic Map