Assay Details
| Gene Symbol: Acss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080575.2 | NP_542142.1 | 2 - 3 | 451 | 63 | |
| GenBank mRNA | AB046742.1 | - | 2 - 3 | 440 | 63 | |
| AK088244.1 | - | 2 - 3 | 430 | 63 | ||
| AK146568.1 | - | 2 - 3 | 452 | 63 | ||
| AK158119.1 | - | 2 - 3 | 446 | 63 | ||
| AK172412.1 | - | 2 - 3 | 437 | 63 | ||
| AK172470.1 | - | 2 - 3 | 426 | 63 | ||
| AK173275.1 | - | 2 - 3 | 412 | 63 | ||
| BC092278.1 | - | 2 - 3 | 480 | 3710505 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acss1
Genomic Map