Assay Details
| Gene Symbol: Acss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080575.2 | NP_542142.1 | 7 - 8 | 1273 | 56 | |
| GenBank mRNA | AB046742.1 | - | 7 - 8 | 1262 | 56 | |
| AK146568.1 | - | 7 - 8 | 1274 | 56 | ||
| AK158119.1 | - | 7 - 8 | 1268 | 56 | ||
| AK173275.1 | - | 7 - 8 | 1234 | 56 | ||
| BC030930.1 | - | 3 - 4 | 447 | 4911679 | 56 | |
| BC092278.1 | - | 7 - 8 | 1302 | 3710505 | 56 | |
More Information
Set Membership: |
|

Acss1
Genomic Map