Assay Details
| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025878.1 | NP_080154.1 | 6 - 7 | 479 | 92 | |
| GenBank mRNA | AB049954.1 | - | 6 - 7 | 503 | 92 | |
| AK002554.1 | - | 3 - 4 | 224 | 92 | ||
| AK010250.1 | - | 6 - 7 | 486 | 92 | ||
| AK076033.1 | - | 3 - 4 | 224 | 92 | ||
| AK076044.1 | - | 3 - 4 | 224 | 92 | ||
| AK088540.1 | - | 6 - 7 | 475 | 92 | ||
| AK166597.1 | - | 6 - 7 | 500 | 92 | ||
| AK167067.1 | - | 6 - 7 | 480 | 92 | ||
| AK168140.1 | - | 6 - 7 | 495 | 92 | ||
| BC021752.1 | - | 6 - 7 | 479 | 5136720 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161697 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161699 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161696 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161695 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161702 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | BC021752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | AK166597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | AK167067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | AK010250.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | AK088540.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | BC021752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | AK166597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | AK167067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | AK010250.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P207964 | AK088540.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139992 | AK168140.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139992 | BC021752.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139992 | AK166597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139992 | AK167067.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139992 | AK010250.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627132 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164373 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | BC021752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | AK166597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | AK167067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | AK010250.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | AK088540.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | BC021752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | AK166597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | AK167067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | AK010250.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P207964 | AK088540.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139992 | AK168140.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139992 | BC021752.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139992 | AK166597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139992 | AK167067.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139992 | AK010250.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | BC021752.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | AK166597.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | AK167067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | AK010250.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | AK088540.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | BC021752.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | AK166597.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | AK167067.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | AK010250.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P207964 | AK088540.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P237281 | AK034502.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31161664 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08769543 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08769534 | - | - |
| Whole Mouse Genome G4122A | A_51_P237281 | AK034502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | BC021752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | AK166597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | AK167067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | AK010250.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | AK088540.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | BC021752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | AK166597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | AK167067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | AK010250.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P207964 | AK088540.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450965 | AK168140.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450965 | BC021752.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450965 | AK166597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450965 | AK167067.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P450965 | AK010250.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134344 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13092637 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13215738 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13215746 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13215754 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13215749 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13215759 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161697 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161696 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161699 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161703 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31161695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31161677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13092635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13215738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13382306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13092648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13343741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13215762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13343744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13215768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13343746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13215772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31161687 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450644 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451164_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450640 | - | - |
| Mouse Genome 430 2.0 Array | 1451164_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451164_PM_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700018963.mm.1 | NM_025878.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018963.mm.1 | NM_025878.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018963.mm.1 | TC1700001987.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018964.mm.1 | NM_025878.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018964.mm.1 | NM_025878.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018964.mm.1 | TC1700001987.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018966.mm.1 | NM_025878.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018966.mm.1 | NM_025878.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018966.mm.1 | TC1700001987.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451164_a_at | - | - |
| Mouse Expression Set 430 | 1451164_PM_a_at | - | - |
| Murine Genome U74v2 Set | 95159_at | - | - |
| Murine Genome U74v2 Set | 95159_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451164_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451164_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451164_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451164_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 104060164 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5570086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570086 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2677047 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2677047 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2443192 | - | - |
| mouse-6 v1.1 (Illumina) | scl0066973.1_60-S | - | - |
| mouse-6 v1.1 (Illumina) | scl50759.5.1_46-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104060164 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066973.1_60-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570086 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104060164 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2865239 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226334 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226334 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2677047 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2865239 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066973.1_60-S | - | - |

Mrps18b
Genomic Map