Assay Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010833.2 | NP_001010833.2 | 11 - 12 | 4611 | 101 | |
| GenBank mRNA | AK129074.1 | - | 5 - 6 | 2347 | 101 | |
| BC085140.1 | - | 6 - 7 | 2480 | 101 | ||
| BC094363.1 | - | 7 - 8 | 3314 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdc1
Genomic Map