Assay Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010833.2 | NP_001010833.2 | 3 - 4 | 702 | 59 | |
| GenBank mRNA | AK033009.1 | - | 3 - 4 | 539 | 59 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mdc1
Genomic Map