Assay Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010833.2 | NP_001010833.2 | 6 - 7 | 2189 | 73 | |
| GenBank mRNA | AK033009.1 | - | 6 - 7 | 2026 | 73 | |
| BC085140.1 | - | 1 - 2 | 58 | 73 | ||
| BC094363.1 | - | 2 - 3 | 892 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mdc1
Genomic Map