Assay Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010833.2 | NP_001010833.2 | 8 - 9 | 2997 | 80 | |
| GenBank mRNA | AK129074.1 | - | 2 - 3 | 733 | 80 | |
| BC085140.1 | - | 3 - 4 | 866 | 80 | ||
| BC094363.1 | - | 4 - 5 | 1700 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mdc1
Genomic Map