Assay Details
| Gene Symbol: Mdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010833.2 | NP_001010833.2 | 9 - 10 | 3035 | 100 | |
| GenBank mRNA | AK129074.1 | - | 3 - 4 | 771 | 100 | |
| BC085140.1 | - | 4 - 5 | 904 | 100 | ||
| BC094363.1 | - | 5 - 6 | 1738 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdc1
Genomic Map