Assay Details
| Gene Symbol: Uqcrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025899.2 | NP_080175.1 | 7 - 8 | 705 | 100 | |
| GenBank mRNA | AK003975.1 | - | 7 - 8 | 679 | 100 | |
| AK004492.1 | - | 7 - 8 | 680 | 100 | ||
| AK005151.1 | - | 7 - 8 | 689 | 100 | ||
| AK007669.1 | - | 6 - 7 | 569 | 100 | ||
| AK075856.1 | - | 7 - 8 | 649 | 100 | ||
| AK077583.1 | - | 7 - 8 | 654 | 100 | ||
| AK088103.1 | - | 7 - 8 | 659 | 100 | ||
| AK167377.1 | - | 7 - 8 | 659 | 100 | ||
| BC003423.1 | - | 7 - 8 | 677 | 2609295 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uqcrc2
Genomic Map