Assay Details
| Gene Symbol: Slx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029420.4 | NP_083696.2 | 3 - 4 | 748 | 101 | |
| NR_033446.1 | 2 - 3 | 696 | 101 | |||
| GenBank mRNA | AK010748.1 | - | 3 - 4 | 748 | 101 | |
| AK010828.1 | - | 3 - 4 | 713 | 101 | ||
| AK049139.1 | - | 3 - 4 | 749 | 101 | ||
| AK053901.1 | - | 2 - 3 | 637 | 101 | ||
| BC055909.1 | - | 3 - 4 | 699 | 101 | ||
| BC145468.1 | - | 2 - 3 | 560 | 101 | ||
| BC145688.1 | - | 3 - 4 | 613 | 40130991 | 101 | |
More Information
Set Membership: |
|

Slx1b
Genomic Map