Assay Details
| Gene Symbol: H2-Q10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010391.4 | NP_034521.1 | 1 - 2 | 91 | 65 | |
| GenBank mRNA | AK131620.1 | - | 1 - 2 | 92 | 65 | |
| BC011215.1 | - | 1 - 2 | 75 | 4196692 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
antigen processing and presentation of peptide antigen via MHC class I
immune response
antigen processing and presentation
receptor binding
beta-2-microglobulin binding
peptide binding
peptide antigen binding
T cell receptor binding
poly(A) RNA binding
TAP binding
protein heterodimerization activity
immune system process
antigen processing and presentation of peptide antigen via MHC class I
immune response
antigen processing and presentation
receptor binding
beta-2-microglobulin binding
peptide binding
peptide antigen binding
T cell receptor binding
poly(A) RNA binding
TAP binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159264 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159261 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995092 | K00614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995092 | NM_010391.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995092 | AK131620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995092 | BC011215.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995092 | BC042572.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037343 | NM_010391.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203866 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995092 | K00614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995092 | NM_010391.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995092 | AK131620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995092 | BC011215.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995092 | BC042572.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037343 | NM_010391.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P496997 | X52490.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P496997 | Y00806.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P496997 | U47325.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P496997 | M37680.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P496997 | M37681.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230267 | K00614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230267 | NM_010391.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230267 | AK131620.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230267 | BC011215.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230267 | BC042572.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230269 | K00614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230269 | NM_010391.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230269 | AK131620.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230269 | BC011215.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P230269 | BC042572.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P428354 | K00614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P428354 | NM_010391.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P428354 | AK131620.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P428354 | BC011215.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P428354 | BC042572.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31159256 | - | - |
| Whole Mouse Genome G4122A | A_51_P230267 | K00614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230267 | NM_010391.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230267 | AK131620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230267 | BC011215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230267 | BC042572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P496997 | X52490.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P496997 | Y00806.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P496997 | U47325.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P496997 | M37680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P496997 | M37681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230269 | K00614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230269 | NM_010391.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230269 | AK131620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230269 | BC011215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P230269 | BC042572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P428354 | K00614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P428354 | NM_010391.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P428354 | AK131620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P428354 | BC011215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P428354 | BC042572.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111989 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13247309 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13247317 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13247301 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13247295 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13247294 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159264 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31159261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31159261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13219971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13139899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13237732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13247301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13154066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13237747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31159305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444844 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425137_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444841 | - | - |
| Mouse Genome 430 2.0 Array | 1425137_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425137_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15007_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15007_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700007058.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007058.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007058.mm.1 | TC1700000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700007067.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007067.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007067.mm.1 | TC1700000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700007061.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007061.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007061.mm.1 | TC1700000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700007054.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007054.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007054.mm.1 | TC1700000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700007056.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007056.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007056.mm.1 | TC1700000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700007060.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007060.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007060.mm.1 | TC1700000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700007065.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007065.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007065.mm.1 | TC1700000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700007066.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007066.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007066.mm.1 | TC1700000688.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700007063.mm.1 | NM_010391.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007063.mm.1 | NM_010391.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700007063.mm.1 | TC1700000688.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425137_PM_a_at | - | - |
| Mouse Expression Set 430 | 1425137_a_at | - | - |
| Murine Genome U74v2 Set | 97174_PM_r_at | - | - |
| Murine Genome U74v2 Set | 101898_s_at | - | - |
| Murine Genome U74v2 Set | 97174_r_at | - | - |
| Murine Genome U74v2 Set | 101898_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425137_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425137_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425137_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425137_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 928591 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3780736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3780736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780736 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2717352 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2717352 | - | - |
| mouse-6 v1.1 (Illumina) | scl50776.2_225-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780736 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50776.2_225-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780736 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2717352 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2717352 | - | - |

H2-Q10
Genomic Map