Assay Details
| Gene Symbol: Zfp668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146259.3 | NP_666371.1 | 2 - 3 | 1449 | 68 | |
| GenBank mRNA | AK047593.1 | - | 3 - 4 | 1489 | 68 | |
| AK053438.1 | - | 2 - 3 | 1450 | 68 | ||
| AK081436.1 | - | 2 - 3 | 1448 | 68 | ||
| AK152456.1 | - | 2 - 3 | 742 | 68 | ||
| AK169556.1 | - | 2 - 3 | 1455 | 68 | ||
| BC028251.1 | - | 2 - 3 | 1812 | 68 | ||
| BC030314.1 | - | 2 - 3 | 1065 | 5371636 | 68 | |
More Information
Set Membership: |
|

Zfp668
Genomic Map