Assay Details
| Gene Symbol: Pnpla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163689.1 | NP_001157161.1 | 5 - 6 | 856 | 63 | |
| NR_028142.1 | 6 - 7 | 938 | 63 | |||
| GenBank mRNA | AK031609.1 | - | 5 - 6 | 814 | 63 | |
| AK076464.1 | - | 5 - 6 | 640 | 63 | ||
| AK085926.1 | - | 5 - 6 | 841 | 63 | ||
| AK150184.1 | - | 5 - 6 | 838 | 63 | ||
| AY510273.1 | - | 5 - 6 | 837 | 63 | ||
| AY731699.1 | - | 5 - 6 | 837 | 63 | ||
| AY894805.1 | - | 5 - 6 | 757 | 63 | ||
| BC019188.1 | - | 6 - 7 | 920 | 5027544 | 63 | |
| BC034190.1 | - | 4 - 5 | 357 | 63 | ||
| BC044737.1 | - | 6 - 7 | 902 | 63 | ||
| BC044781.2 | - | 6 - 7 | 902 | 5151911 | 63 | |
| BC054825.1 | - | 6 - 7 | 818 | 63 | ||
| BC064747.1 | - | 5 - 6 | 828 | 30024535 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnpla2
Genomic Map