Assay Details
| Gene Symbol: Entpd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172117.5 | NP_742115.2 | 1 - 2 | 157 | 132 | |
| GenBank mRNA | AK030789.1 | - | 1 - 2 | 158 | 132 | |
| BC038126.1 | - | 1 - 2 | 146 | 1348160 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC038126.1, NM_172117.5 transcripts.
Set Membership: |
|

Entpd6
Genomic Map