Assay Details
| Gene Symbol: 3300002I08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027017.1 | NP_081293.4 | 1 - 2 | 181 | 136 | |
| GenBank mRNA | AB470968.1 | - | 1 - 2 | 141 | 136 | |
| AK013537.1 | - | 1 - 2 | 160 | 136 | ||
| AK014369.1 | - | 1 - 2 | 181 | 136 | ||
| BC132262.1 | - | 1 - 2 | 122 | 40130539 | 136 | |
| BC132264.1 | - | 1 - 2 | 122 | 40130541 | 136 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB470968.1, BC132262.1, BC132264.1, NM_027017.1 transcripts.
Set Membership: |
|

3300002I08Rik
Genomic Map