Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110209.1 | NP_001103679.1 | 11 - 12 | 1056 | 97 | |
| NM_027133.3 | NP_081409.1 | 11 - 12 | 1056 | 97 | ||
| GenBank mRNA | AK014897.1 | - | 11 - 12 | 953 | 97 | |
| AK048501.1 | - | 2 - 3 | 1299 | 97 | ||
| AK082900.1 | - | 10 - 11 | 981 | 97 | ||
| AK136506.1 | - | 11 - 12 | 1031 | 97 | ||
| AK137445.1 | - | 11 - 12 | 1057 | 97 | ||
| AK173241.1 | - | 11 - 12 | 988 | 97 | ||
| AY312281.1 | - | 11 - 12 | 977 | 97 | ||
| BC057961.1 | - | 11 - 12 | 983 | 5353629 | 97 | |
| BC060153.1 | - | 10 - 11 | 895 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lnp
Genomic Map