Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110209.1 | NP_001103679.1 | 2 - 3 | 224 | 148 | |
| NM_027133.3 | NP_081409.1 | 2 - 3 | 224 | 148 | ||
| GenBank mRNA | AK009311.1 | - | 2 - 3 | 198 | 148 | |
| AK014897.1 | - | 2 - 3 | 122 | 148 | ||
| AK053593.1 | - | 2 - 3 | 193 | 148 | ||
| AK082900.1 | - | 2 - 3 | 208 | 148 | ||
| AK137445.1 | - | 2 - 3 | 225 | 148 | ||
| AK143497.1 | - | 2 - 3 | 193 | 148 | ||
| AK173241.1 | - | 2 - 3 | 156 | 148 | ||
| AY312281.1 | - | 2 - 3 | 145 | 148 | ||
| BC057961.1 | - | 2 - 3 | 151 | 5353629 | 148 | |
| BC060153.1 | - | 1 - 2 | 62 | 149 | ||
| BY161702.1 | - | 2 - 3 | 148 | |||
| BY770278.1 | - | 2 - 3 | 224 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK143497.1, AY312281.1, BC057961.1, NM_001110209.1, NM_027133.3 transcripts.
Set Membership: |
|

Lnp
Genomic Map